missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLC-gamma 1 Antibody (2F3D11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | PLC-gamma 1 |
|---|---|
| Clone | 2F3D11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18208881
|
Novus Biologicals
NBP2-61790 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698947
|
Novus Biologicals
NBP2-61790-0.025ml |
0.025 mL |
£161.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PLC-gamma 1 Monoclonal specifically detects PLC-gamma 1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| PLC-gamma 1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| 5335 | |
| Purified recombinant fragment of human PLCG1 (AA: 39-181) expressed in E. Coli. | |
| Primary | |
| 89.8 kDa |
| 2F3D11 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| Lipid and Metabolism, Phospho Specific, Signal Transduction | |
| EC 3.1.4.11, gamma 1 (formerly subtype 148), monophosphatidylinositol phosphodiesterase, NCKAP3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1, phosphatidylinositol phospholipase C, phosphoinositidase C, phosphoinositide phospholipase C, Phosphoinositide phospholipase C-gamma-1, phospholipase C, gamma 1, phospholipase C-148, Phospholipase C-gamma-1, Phospholipase C-II, PLC11-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1, PLC148, PLC-148, PLC-gamma-1, PLC-II1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, triphosphoinositide phosphodiesterase | |
| PLCG1 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title