missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLC-delta 3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP3-20228
This item is not returnable.
View return policy
Description
PLC-delta 3 Polyclonal antibody specifically detects PLC-delta 3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Specifications
| PLC-delta 3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/ml | |
| Western Blot 1-3 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 6 ug/ml, Peptide ELISA Detection limit 1:32000 | |
| EC 3.1.4.11, KIAA1964, MGC71172, Phosphoinositide phospholipase C-delta-3, phospholipase C, delta 3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3, Phospholipase C-delta-3, PLC delta3, PLC-delta-3 | |
| Peptide with sequence C-HQYSGEDRVLSAPE, from the internal region of the protein sequence according to NP_588614.1. | |
| 0.1 mg | |
| Lipid and Metabolism | |
| 113026 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction