missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLC-beta 3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93347-0.1ml
This item is not returnable.
View return policy
Description
PLC-beta 3 Polyclonal antibody specifically detects PLC-beta 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PLC-beta 3 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3, EC 3.1.4.11, FLJ37084, Phosphoinositide phospholipase C-beta-3, phospholipase C, beta 3 (phosphatidylinositol-specific), Phospholipase C-beta-3, PLC beta 3, PLC-beta-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1010-1234 of human PLC-beta 3 (NP_001303243.1). GALGGAADVEDTKEGEDEAKRYQEFQNRQVQSLLELREAQVDAEAQRRLEHLRQALQRLREVVLDANTTQFKRLKEMNEREKKELQKILDRKRHNSISEAKMRDKHKKEAELTEINRRHITESVNSIRRLEEAQKQRHDRLVAGQQQVLQQLAEEEPKLLAQLAQECQEQRARLPQEIRRSLLGEMPEGLGDGPLVACASNGHAPGSSGHLSGADSESQEENTQL | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5331 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction