missing translation for 'onlineSavingsMsg'
Learn More

PLC-beta 1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93680-0.1ml

 View more versions of this product

Product Code. 18620681

  • £366.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PLC-beta 1 Polyclonal antibody specifically detects PLC-beta 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLC-beta 1
Polyclonal
Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
EC 3.1.4.11, EIEE12, FLJ45792, inositoltrisphosphohydrolase, KIAA05811-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta 1, monophosphatidylinositol phosphodiesterase, phosphoinositidase C, Phosphoinositide phospholipase C-beta-1, phospholipase C, beta 1 (phosphoinositide-specific), Phospholipase C-beta-1, Phospholipase C-I, PI-PLC, PLC-154, PLC154,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-1, PLCB1A, PLCB1B, PLC-beta-1, PLC-I1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, triphosphoinositide phosphodiesterase
Recombinant fusion protein containing a sequence corresponding to amino acids 917-1216 of human PLCB1 (NP_056007.1). KSFVKLQKKHYKEMKDLVKRHHKKTTDLIKEHTTKYNEIQNDYLRRRAALEKSAKKDSKKKSEPSSPDHGSSTIEQDLAALDAEMTQKLIDLKDKQQQQLLNLRQEQYYSEKYQKREHIKLLIQKLTDVAEECQNNQLKKLKEICEKEKKELKKKMDKKRQEKITEAKSKDKSQMEEEKTEMIRSYIQEVVQYIKRLEEAQSKRQEKLVEKHKEIRQQILDEKPKLQVELEQEYQDKFKRLPLEILEFVQEAMKGKISEDSNHGSAPLSLSSDPGKVNHKTPSSEELGGDIPGKEFDTPL
0.1 mL
Lipid and Metabolism, Wnt Signaling Pathway
23236
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.