missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLBD2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06057APC
This item is not returnable.
View return policy
Description
PLBD2 Polyclonal antibody specifically detects PLBD2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| PLBD2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| LAMA-like protein 2,76 kDa protein, lamina ancestor homolog 2, mannose-6-phosphate protein associated protein p76, P76, phospholipase B domain containing 2, phospholipase B domain-containing protein 2, phospholipase B-like 2 32 kDa form, phospholipase B-like 2 45 kDa form, PLB homolog 2, putative phospholipase B-like 2 | |
| Produced in rabbits immunized with E. coli-derived Human PLBD2 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 196463 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction