missing translation for 'onlineSavingsMsg'
Learn More

PLBD2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18370806 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18370806 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18370806 Supplier Novus Biologicals Supplier No. NBP306057AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLBD2 Polyclonal specifically detects PLBD2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLBD2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias LAMA-like protein 2,76 kDa protein, lamina ancestor homolog 2, mannose-6-phosphate protein associated protein p76, P76, phospholipase B domain containing 2, phospholipase B domain-containing protein 2, phospholipase B-like 2 32 kDa form, phospholipase B-like 2 45 kDa form, PLB homolog 2, putative phospholipase B-like 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human PLBD2 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196463
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.