missing translation for 'onlineSavingsMsg'
Learn More

Platinum wire, 0.5mm (0.02in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Catalog Number 11349418
Click to view available options
Quantity:
25 cm
100 cm
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]
7440-06-4
Pt
195.08
MFCD00011179
BASFCYQUMIYNBI-UHFFFAOYSA-N
black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
23939
CHEBI:33400
platinum
[Pt]

Platinum wire; 99.95% metal basis; Alfa Aesar; 0.5mm diameter; annelaled; 25cm length; 4.21g/m, Temper soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99.95% (metals basis)
Quantity 25 cm
Chemical Name or Material Platinum wire

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'