missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum, Technical, Metal Wire, 0.5mm, SLR, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 cm
50 cm
100 cm
Unit Size:
100cm
10cm
50cm
Specifications
Specifications
| Color | Silver |
| CAS | 7740-06-4 |
| Melting Point | 1772°C |
| Boiling Point | 3827°C |
| MDL Number | 11179 |
| Quantity | 50 cm |
| Formula Weight | 195.08g/mol |
| Packaging | Plastic powder jar |
| Form | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction