missing translation for 'onlineSavingsMsg'
Learn More

Platinum, Technical, Metal Wire, 0.5mm, SLR, Fisher Chemical™

Product Code. 11468752 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 cm
50 cm
100 cm
Unit Size:
100cm
10cm
50cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11468752 100 cm 100cm
10477260 10 cm 10cm
10191430 50 cm 50cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468752 Supplier Fisher Chemical Supplier No. P/3640/87
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula Pt
Molecular Weight (g/mol) 195.08
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Silver
CAS 6-4-7440
Melting Point 1772°C
Boiling Point 3827°C
Quantity 100 cm
Packaging Plastic powder jar
Form Solid
MDL Number MFCD00011179
Formula Weight 195.08g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.