missing translation for 'onlineSavingsMsg'
Learn More

Platinum, Technical, Metal Wire, 0.5mm, SLR, Fisher Chemical™

Product Code. 10477260 Shop All Fisher Chemical Products
Click to view available options
Quantity:
10 cm
50 cm
100 cm
Unit Size:
100cm
10cm
50cm
This item is not returnable. View return policy

Product Code. 10477260

Brand: Fisher Chemical P/3640/88

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula Pt
Molecular Weight (g/mol) 195.08
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Color Silver
CAS 6-4-7440
Melting Point 1772°C
Boiling Point 3827°C
Quantity 10 cm
Packaging Plastic powder jar
Form Solid
MDL Number MFCD00011179
Formula Weight 195.08g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.