missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Platinum Taq Monoclonal Antibody
GREENER_CHOICE

Product Code. 10123812
Change view
Click to view available options
Quantity:
250 units
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10123812 250 units Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10123812 Supplier Invitrogen™ Supplier No. 10965028

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Monoclonal Antibody

One unit of Taq DNA polymerase incorporates 10 nmol of deoxyribonucleotide into DNA in 30 minutes at 74°C. One unit is defined as the amount of product required to inhibit one unit of Taq DNA polymerase. PCR Functional Assay Specifications: PCR reactions containing 100 ng, 10 ng, and 5 ng of input human genomic DNA yield 40 ng, 10 ng, and 5 ng of an 800 bp target, respectively, when observed on an ethidium bromide stained gel. In a radioactive nucleotide incorporation assay, Taq DNA polymerase activity has been successfully inhibited by 10965-010.

Platinum™ Taq Antibody is a proprietary inhibitor which, when bound to Taq DNA polymerase, inactivates polymerase activity. This reagent provides an automatic 'hot start' for Taq DNA polymerase in PCR. Hot starts are typically used in PCR to increase sensitivity, specificity, and yield while allowing assembly of reactions at ambient temperatures. The extra time, effort, and contamination risks associated with manual hot start procedures are addressed with the use of Platinum™ Taq Antibody.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Platinum Taq
Applications Functional Assay, Inhibition Assays
Classification Monoclonal
Conjugate Unconjugated
Formulation TBS with 2mM Na2PO4, 50% glycerol, 0.1mM EDTA, 1mM DTT and no preservative; pH 8
Gene Alias Taq DNA polymerase
Host Species Not Available
Immunogen Taq DNA Polymerase.
Purification Method Purified
Quantity 250 units
Regulatory Status RUO
Primary or Secondary Primary
Target Species Tag
Content And Storage -20°C
Product Type Antibody
Form Liquid
Shipping Condition Wet or Dry Ice
Target Molecule Tag
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.