missing translation for 'onlineSavingsMsg'
Learn More

Platinum rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)

Product Code. 11387397
Change view
Click to view available options
Quantity:
10 mm
Unit Size:
10mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387397 10 mm 10mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11387397 Supplier Thermo Scientific Alfa Aesar Supplier No. 012556.HF
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7440-06-4 | Pt | 195.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6-4-7440
Quantity 10 mm
Odor Odorless
Weight ≈6.8g/cm
Form Rod
Molecular Formula Pt
MDL Number MFCD00011179
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES [Pt]
IUPAC Name platinum
Molecular Weight (g/mol) 195.08
PubChem CID 23939
ChEBI CHEBI:33400
Percent Purity 99.99%
Grade Premion
Assay (metals basis)
Chemical Name or Material Platinum rod, 6.35mm (0.25 in.) dia.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.