Learn More
Platinum Rhodium Palladium gauze, 80 mesh knitted from 0.076mm (0.003in) dia wire, 99.9% (metals basis), Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 012216.FF
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Platinum Rhodium Palladium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis) is used as a catalyst in the preparation of nitric acid from ammonia. It is also used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum Rhodium Palladium gauze | |
Pt:Rh:Pd; 90:5:5 wt% | |
MFCD00011179 | |
Insoluble in water. | |
80 Mesh |
Woven from 0.076mm (0.003 in.) dia. wire | |
(metals basis) | |
25 x 25 mm | |
99.9% | |
≈0.39g/25x25mm |
RUO – Research Use Only