Learn More
Platinum Rhodium Palladium gauze, 80 mesh knitted from 0.076mm (0.003in) dia wire, 99.9% (metals basis)
Description
Platinum Rhodium Palladium gauze, 80 mesh woven from 0.076mm (0.003in) dia wire, 99.9% (metals basis) is used as a catalyst in the preparation of nitric acid from ammonia. It is also used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Platinum Rhodium Palladium gauze |
| Name Note | Woven from 0.076mm (0.003 in.) dia. wire |
| Avg. Mol. Wt. or Mol. Wt. Range | Pt:Rh:Pd; 90:5:5 wt% |
| Quantity | 100 x 100 mm |
| Mesh Size | 80 Mesh |
| Weight | ≈0.39g/25x25mm |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD00011179 |
| Solubility Information | Insoluble in water. |
| Percent Purity | 99.9% |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.