Learn More
Platinum Rhodium foil, 0.05mm (0.002in) thick, 99.9% (metals basis)
Description
Platinum Rhodium foil, 0.05mm (0.002in) thick, 99.9% (metals basis) is used in the glass industry for furnace linings and complex fabrications. It is also used as a plating material. Further, it acts as a catalyst and as a standard for calibration of base metal thermocouples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Platinum Rhodium foil |
| Name Note | 0.05mm (0.002 in.) thick |
| Avg. Mol. Wt. or Mol. Wt. Range | Pt:Rh; 90:10 wt% |
| Quantity | 100 x 100 mm |
| Odor | Odorless |
| Weight | ≈0.62g/25x25mm |
| Form | Foil |
| Assay Percent Notes | (metals basis) |
| MDL Number | MFCD00198186 |
| Solubility Information | Insoluble in water. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.