missing translation for 'onlineSavingsMsg'
Learn More

Platinum Rhodium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)

Product Code. 11384767
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
25mm
Each
This item is not returnable. View return policy

Product Code. 11384767

Brand: Thermo Scientific Alfa Aesar 041811.FF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used as a standard for calibration of base metal thermocouples. It is also used in the glass industry for furnace linings and complex fabrications. | Pt:Rh; 80:20 wt%

Platinum Rhodium foil is used as a standard for calibration of base metal thermocouples. It is also used in the glass industry for furnace linings and complex fabrications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Rhodium foil
Quantity 25 x 25 mm
Molecular Formula Pt:Rh; 80:20 wt%
MDL Number MFCD00198186
Solubility Information Insoluble in water.
Percent Purity 99.9%
Packaging Tube

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.