missing translation for 'onlineSavingsMsg'
Learn More

Platinum Rhodium foil, 0.025mm (0.001in) thick, 99.9% (metals basis)

Product Code. 11335939
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
25mm
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11335939 50 x 50 mm Each
11334767 25 x 25 mm 25mm
11324767 100 x 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11335939 Supplier Thermo Scientific Alfa Aesar Supplier No. 041808.FI
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Platinum Rhodium foil, 0.025mm (0.001in) thick, 99.9% (metals basis) is used as a standard for calibration of base metal thermocouples. It is also used in the glass industry for furnace linings and complex fabrications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Rhodium foil
Name Note 0.025mm (0.001 in.) thick
Avg. Mol. Wt. or Mol. Wt. Range Pt:Rh; 90:10 wt%
Quantity 50 x 50 mm
Odor Odorless
Weight ≈0.31g/25x25mm
Form Foil
Assay Percent Notes (metals basis)
MDL Number MFCD00198186
Solubility Information Insoluble in water.
Percent Purity 99.9%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.