missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human 13-Plex Human ProcartaPlex™ Panel 2, Platinum

Product Code. 15582537
Click to view available options
Format:
Preconfigured Panel
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 15582537

Brand: Invitrogen™ EPXP13010100901

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • Platinum Human Standard Mix P1
  • Platinum Human Control P1 High
  • Platinum Human Control P1 Low
  • Platinum Human Panel 2 Bead Mix
  • Platinum Human Panel 2 Detection Antibody Mix
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Assay Diluent for Plasma Samples
  • Assay Diluent for Serum Samples
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well

Bead-based multiplexing assay system for quantificaton of multiple protein biomarkers in a single sample. Newly developed matrix-type-specific diluents for plasma and serum overcome matrix effects and assure high performance specifications (e.g. spike and dilution recovery results 70-130%)

  • Platinum ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • Platinum ProcartaPlex Panels are ready-to-use kits and are not combinable with other Platinum ProcartaPlex Simplex kits or kits from different format
TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Species Human
Detection Method Fluorescence
Conjugate R-PE
Bead Type See product documentation
Assay Sensitivity See product documentation
Format Preconfigured Panel
For Use With (Equipment) Luminex Instruments
Product Line ProcartaPlex
Sample Type Serum, Plasma
Sample Volume Serum, Plasma: 25 μL
Content And Storage 2 vials Platinum Human Standard Mix P1
2 vials Platinum Human Control P1 High
2 vials Platinum Human Control P1 Low
1 vial Platinum Human Panel 2 Bead Mix
1 vial Platinum Human Panel 2 Detection Antibody Mix
1 bottle Reading Buffer
1 bottle Wash Buffer 10x
1 bottle Streptavidin-PE
1 bottle Assay Diluent for Plasma Samples
1 bottle Assay Diluent for Serum Samples
PCR 8-Tube Strip
Adhesive Film
Greiner Microplate black 96 well

Store at 2-8°C.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.