missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum, nominally 70% on high surface area advanced carbon support, HiSPEC™ 13100, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | Gray to Black |
Odor | Odorless |
Quantity | 5 g |
UN Number | UN1325 |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Platinum, HiSPEC™ 13100, Nominally 70% on high surface area advanced carbon support |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'