missing translation for 'onlineSavingsMsg'
Learn More

Platinum, nominally 60% on high surface area advanced carbon support, HiSPEC™ 9100, Thermo Scientific Chemicals

 

£191.00 - £781.33

Chemical Identifiers

CAS 7440-06-4
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platine, platino, on carbon, platinum, metal, platin german, platinum, elemental, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11439960
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044728.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11449960
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044728.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Platinum, nominally 60% on high surface area advanced carbon support, HiSPEC^t 9100 is used for anodes and cathodes in proton exchange membrane fuel cells (PEMFC) and direct methanol fuel cells (DMFC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
Specifications

Chemical Identifiers

7440-06-4
195.08
BASFCYQUMIYNBI-UHFFFAOYSA-N
23939
platinum
Pt
MFCD00011179
black, platin, sponge, platine, platino, on carbon, platinum, metal, platin german, platinum, elemental, iv ion
CHEBI:33400
[Pt]

Specifications

7440-06-4
Pt
UN1325
BASFCYQUMIYNBI-UHFFFAOYSA-N
platinum
23939
Platinum, HiSPEC™ 9100, Nominally 60% on high surface area advanced carbon support
Odorless
MFCD00011179
black, platin, sponge, platine, platino, on carbon, platinum, metal, platin german, platinum, elemental, iv ion
[Pt]
195.08
CHEBI:33400
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only