missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum, nominally 60% on high surface area advanced carbon support, HiSPEC™ 9100, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Platinum, nominally 60% on high surface area advanced carbon support, HiSPEC^t 9100 is used for anodes and cathodes in proton exchange membrane fuel cells (PEMFC) and direct methanol fuel cells (DMFC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Odor | Odorless |
Quantity | 5 g |
UN Number | UN1325 |
Chemical Name or Material | Platinum, HiSPEC™ 9100, Nominally 60% on high surface area advanced carbon support |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'