missing translation for 'onlineSavingsMsg'
Learn More

Platinum (IV) Oxide, TRC

Product Code. 30412515
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30412515 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30412515 Supplier Toronto Research Chemicals Supplier No. TRCP5780005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum(IV) oxide
CAS 1314-15-4
Quantity 5 g
Molecular Formula O2 Pt
Synonym Platinum dioxide
InChI Formula InChI=1S/2O.Pt
SMILES O=[Pt]=O
IUPAC Name dioxoplatinum
Molecular Weight (g/mol) 227.0768
Formula Weight 226.9546
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.