missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis)

Product Code. 10249603 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10249603 5 g Glass bottle 5g
10773211 250 mg Glass bottle 250mg
10276600 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10249603 Supplier Thermo Scientific Acros Supplier No. 211930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 12137-21-2 | H2O3Pt | 245.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12137-21-2
Molecular Formula H2O3Pt
Molecular Weight (g/mol) 245.10
MDL Number MFCD00066964
InChI Key SPGAMGILENUIOF-UHFFFAOYSA-N
Synonym dioxoplatinum hydrate, platinum iv oxide monohydrate, platinum dioxide hydrate, platinum iv oxide hydrate, platinoxydhydrat, acmc-1bujp, platinum 1v oxide hydrate, pto2 h2o
PubChem CID 11139377
SMILES O.O=[Pt]=O

Specifications

Color Brown to Black
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.8%
Linear Formula PtO2·H2O
Fieser 01,890
Merck Index 10, 7398
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in conc.acids and in dil.sol. of koh
IUPAC Name dioxoplatinum hydrate
Formula Weight 245.1
Percent Purity 99.80%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Platinum(IV) oxide monohydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Platinum(IV) oxide monohydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.