missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Pt 84.4% min

Product Code. 11363998
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363998 1 g 1g
11343998 0.25 g 0.25g
11353998 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363998 Supplier Thermo Scientific Alfa Aesar Supplier No. 040402.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 1314-15-4 | O2Pt | 227.08 g/mol

Platinum(IV) oxide is used as catalyst for hydrogenation reactions. It is applied in the hydrogenation of alkenes to alkanes, nitro compounds to amines and ketones to alcohols. It is also involved in the hydrogenolysis of cyclopropyl group in to an isopropyl group and in selective oxidation of primary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-15-4
Molecular Formula O2Pt
Molecular Weight (g/mol) 227.08
MDL Number MFCD00011184
InChI Key YKIOKAURTKXMSB-UHFFFAOYSA-N
IUPAC Name dioxoplatinum
SMILES O=[Pt]=O

Specifications

Melting Point 450°C
Color Brown
Physical Form Powder
Quantity 1 g
Linear Formula PtO2
UN Number UN1479
Merck Index 14,7527
Solubility Information Soluble in caustic potash solution. Insoluble in water,acid,aquaregia.
Formula Weight 227.09
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 84.4% min.
Odor Odorless
Density 10.2 g/mL
Assay (metals basis)
Chemical Name or Material Platinum(IV) oxide, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Platinum(IV) oxide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Platinum oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.