missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) nitrate, solution, Pt 15% w/w

Product Code. p-7032997
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 15407817

Brand: Thermo Scientific Alfa Aesar H37737.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10102-09-7 | N4O12Pt

Platinum(IV) nitrate acts as a precursor in the production of platinum dioxide on support surface.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-09-7
Molecular Formula N4O12Pt
MDL Number MFCD25372548
Synonym platinum 4+ ion nitric acid

Specifications

UN Number UN3093
Solubility Information Miscible with water.
Formula Weight 443.1
Odor Acidic
Quantity 1 g
Assay Pt 15% w/w
Chemical Name or Material Platinum(IV) nitrate, solution
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Platinum(IV) nitrate, solution, Pt 15% w/w
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P314-Get medical advice/attention if you feel unwell.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Platinum(IV) nitrate, Nitric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.