missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) iodide, 99.95% (Metals basis), Pt 27.3% min., Thermo Scientific Chemicals, Premion™

Product Code. 11323998
Change view
Click to view available options
Quantity:
0.25 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323998 5 g 5g
11313998 0.25 g 0.25g
11333998 5 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11323998 Supplier Thermo Scientific Alfa Aesar Supplier No. 040401.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-46-7
Molecular Formula I4Pt
MDL Number MFCD00016292

Specifications

Melting Point 130°C (decomposition)
Color Black
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.95% (metals basis)
Solubility Information Decomposes in water. Soluble in alcohol,acetone,alkali,HI,KI,liquid NH3
Formula Weight 702.7
Appearance Black powder
Density 6.06
Chemical Name or Material Platinum(IV) iodide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.