missing translation for 'onlineSavingsMsg'
Learn More

Platinum(Iv) Dioxide, Pure, Fisher Chemical

Product Code. 10599000 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Vial
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10599000 1 g Vial 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10599000 Supplier Fisher Chemical Supplier No. P/3652/43
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 450°C
CAS 1314-15-4
Color Brown
Physical Form Powder/Solid
Quantity 1 g
Molecular Formula O2Pt
MDL Number MFCD00011184
InChI Key YKIOKAURTKXMSB-UHFFFAOYSA-N
SMILES O=[Pt]=O
IUPAC Name dioxoplatinum
Molecular Weight (g/mol) 227.08
Formula Weight 227.09g/mol
Packaging Vial
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Platinum(IV) oxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.