missing translation for 'onlineSavingsMsg'
Learn More

Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min

Product Code. 11330298
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11330298

Brand: Thermo Scientific Alfa Aesar 043703.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in electroplating, as a catalyst and analytical reagent. Also used in the production of semiconductor epitaxy, diffusion process. | CAS: 13454-96-1 | Cl4Pt | 336.88 g/mol

Platinum(IV) chloride is used in electroplating, as a catalyst and analytical reagent. Also used in the production of semiconductor epitaxy, diffusion process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13454-96-1
Molecular Formula Cl4Pt
Molecular Weight (g/mol) 336.88
MDL Number MFCD00011182
InChI Key FBEIPJNQGITEBL-UHFFFAOYSA-J
Synonym platinum tetrachloride, platinum iv chloride, platinum iv tetrachloride, platinum chloride ptcl4, platinum chloride ptcl4 , sp-4-1, platinum chloride ptcl4 6ci,7ci,8ci, platinum iv chloride, premion, ptcl4, platinumchloride ptcl4
PubChem CID 26031
IUPAC Name tetrachloroplatinum
SMILES Cl[Pt](Cl)(Cl)Cl

Specifications

Melting Point 370°C (dec.)
Physical Form Crystalline
Quantity 5 g
UN Number UN2923
Solubility Information Soluble in water ( 587g/L ),acetone,slightly soluble in ethanol,ammonia. Insoluble in ether.
Formula Weight 336.9
Percent Purity ≥99.99%
Packaging Glass bottle
Sensitivity Hygroscopic
Specific Gravity 4.303
Density 4.303
Chemical Name or Material Platinum(IV) chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.