missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum(IV) chloride, Premion™, 99.99+% (metals basis), Pt 57% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Platinum(IV) chloride is used as a catalyst and analytical reagent. Further, it is used in the production of semiconductor epitaxy and in electroplating process. It is involved in the preparation of platinum(II) chloride .
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 370°C (decomposition) |
Color | Brown |
Physical Form | Crystalline Soluble |
Quantity | 5 g |
UN Number | UN2923 |
Solubility Information | Soluble in water and acetone. Slightly soluble in ethanol and insoluble in ether. |
Formula Weight | 336.9 |
Percent Purity | ≥99.99% |
Grade | Premion™ |
Concentration or Composition (by Analyte or Components) | Pt 57% min |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'