missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 1.0mm (0.04in) dia

Product Code. 11385407
Change view
Click to view available options
Quantity:
5 cm
25 cm
100 cm
Unit Size:
100cm
25cm
5cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11385407 25 cm 25cm
11395407 5 cm 5cm
11305417 100 cm 100cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11385407 Supplier Thermo Scientific Alfa Aesar Supplier No. 042678.BU
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Iridium wire
Name Note 1.0mm (0.04 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 90:10 wt%
Quantity 25 cm
Odor Odorless
Weight ≈0.85g/5cm
Form Wire
MDL Number MFCD02091683

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.