missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.5mm (0.02in) dia, hard, Thermo Scientific Chemicals

Brand:  Thermo Scientific Alfa Aesar 010054.BS

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11386757

  • £118.00 / 10cm
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Fabrication of metal microelectrodes for electrical stimulation of nervous tissue and electrophysiological recordings. Inoculating needles and leadwires for implants. Electrode material for gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Platinum Iridium wire
Pt:Ir; 80:20 wt%
10 cm
Odorless
Wire
0.5mm (0.02 in.) dia., Hard, Temper: as drawn
MFCD02091683
Insoluble in water
≈0.44g/10cm
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only