missing translation for 'onlineSavingsMsg'
Learn More

Platinum Iridium wire, 0.152mm (0.006in) dia, hard, 99.9% (metals basis excluding precious metals)

Product Code. 11349498
Change view
Click to view available options
Quantity:
10 cm
50 cm
Unit Size:
10cm
50cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349498 10 cm 10cm
11339498 50 cm 50cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349498 Supplier Thermo Scientific Alfa Aesar Supplier No. 043331.BS
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum Iridium wire
Name Note 0.152mm (0.006 in.) dia., Hard
Avg. Mol. Wt. or Mol. Wt. Range Pt:Ir; 80:20 wt%
Quantity 10 cm
Odor Odorless
Weight ≈0.04g/10cm
Form Wire
Assay Percent Notes (metals basis excluding precious metals)
MDL Number MFCD02091683
Percent Purity 99.9%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.