missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) chloride, 73% Pt

Product Code. 10267593 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10267593 250 mg Glass bottle 250mg
10318050 1 g Glass bottle 1g
10042590 5 g Glass bottle 5g
11954191 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10267593 Supplier Thermo Scientific Acros Supplier No. 195382500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10025-65-7 | Cl2Pt | 265.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-65-7
Molecular Formula Cl2Pt
Molecular Weight (g/mol) 265.98
MDL Number MFCD00011181
InChI Key CLSUSRZJUQMOHH-UHFFFAOYSA-L
Synonym Platinous chloride
IUPAC Name platinum(2+) dichloride
SMILES [Cl-].[Cl-].[Pt++]

Specifications

Melting Point 581.0°C
Color Brown to Gray-Green
Physical Form Powder
Quantity 250 mg
Linear Formula PtCl2
Fieser 02,272
Merck Index 15,7643
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid,insoluble in alcohol and ether
Formula Weight 266
Percent Purity 73% (Pt)
Packaging Glass bottle
Specific Gravity 6.05
Density 6.0500g/mL
Chemical Name or Material Platinum(II) chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Platinum(II) chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.