missing translation for 'onlineSavingsMsg'
Learn More

Platinum(II) chloride, 98%

Product Code. 11487867
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487867 1 g 1g
11497867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487867 Supplier Thermo Scientific Alfa Aesar Supplier No. A16549.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 10025-65-7 | Cl2Pt | 265.98 g/mol

Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond, and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum(II) chloride is used as a catalyst for a variety of C-C bond, C-O bond, and C-N bond formation reactions. It acts as a precursor to prepare other platinum compounds such as cisplatin and dichloro(cycloocta-1,5-diene)platinum(II).

Solubility
Soluble in ammonia and hydrochloric acid. Insoluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-65-7
Molecular Formula Cl2Pt
Molecular Weight (g/mol) 265.98
MDL Number MFCD00011181
InChI Key CLSUSRZJUQMOHH-UHFFFAOYSA-L
IUPAC Name platinum(2+) dichloride
SMILES [Cl-].[Cl-].[Pt++]

Specifications

Melting Point 581°C (decomposition)
Quantity 1 g
Linear Formula PtCl2
UN Number UN3260
Merck Index 14,7528
Solubility Information Soluble in ammonia and hydrochloric acid. Insoluble in water,alcohol and ether.
Formula Weight 266
Percent Purity 98%
Density 5.87
Chemical Name or Material Platinum(II) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.