missing translation for 'onlineSavingsMsg'
Learn More

Platinum foil, 0.25mm (0.01in) thick, 99.9% (metals basis)

Product Code. 11375719
Click to view available options
Quantity:
50 x 50 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11375719

Brand: Thermo Scientific Alfa Aesar 000861.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 7440-06-4

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6-4-7440
Quantity 50 x 50 mm
Percent Purity 99.9%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.