missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Platinum chloride, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Platinum chloride |
| CAS | 10025-65-7 |
| Molecular Formula | Cl2Pt |
| Quantity | 250 mg |
| Synonym | Platinous chloride, Platinous dichloride, Platinum dichloride, Platinum(II) chloride, Platinum(II) dichloride |
| InChI Formula | InChI=1S/2ClH.Pt/h2*1H;/q;;+2/p-2 |
| SMILES | [Cl-].[Cl-].[Pt+2] |
| IUPAC Name | platinum(2+);dichloride |
| Molecular Weight (g/mol) | 265.99 |
| Formula Weight | 264.9025 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction