missing translation for 'onlineSavingsMsg'
Learn More

Platinum chloride, TRC

Product Code. 30425058
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 30425058

Brand: Toronto Research Chemicals TRCP578100500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum chloride
CAS 10025-65-7
Molecular Formula Cl2Pt
Quantity 500 mg
Synonym Platinous chloride, Platinous dichloride, Platinum dichloride, Platinum(II) chloride, Platinum(II) dichloride
InChI Formula InChI=1S/2ClH.Pt/h2*1H;/q;;+2/p-2
SMILES [Cl-].[Cl-].[Pt+2]
IUPAC Name platinum(2+);dichloride
Molecular Weight (g/mol) 265.99
Formula Weight 264.9025
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.