missing translation for 'onlineSavingsMsg'
Learn More

Platinum, AAS standard solution, Specpure™ Pt 1000μg/mL, Thermo Scientific Chemicals

Product Code. 11339179
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: Liquid

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-6-6
Molecular Formula Pt in 20% HCl
MDL Number MFCD00011179

Specifications

Quantity 100 mL
Assay Percent Range Pt 1000μg/mL
Odor Characteristic
Density Approx. 1g/cm3 at 20°C
Physical Form Liquid
Chemical Name or Material Platinum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'