missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 5% on carbon, dry

Product Code. 11429133
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429133 5 g 5g
11339216 1 g 1g
11439133 25 g 25g
11329216 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11429133 Supplier Thermo Scientific Alfa Aesar Supplier No. A11186.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-06-4 | Pt | 195.08 g/mol

Platinum is a catalyst used for hydrgenation of alkenes, alkynes, nitro groups; also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Platinum is a catalyst used for hydrgenation of alkenes, alkynes, nitro groups; also used for hydrogenolysis of cyclopropanes, catalyzes oxidation of alcohols.

Solubility
Soluble in boiling aqua regia (reacts), and molten alkali cyanides (reacts). Insoluble in water, mineral acids, and organic acids.

Notes
Do not store together with acids. Incompatible with Strong acids, strong oxidizing agents & organics Solvent vapors. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-4-7440
Molecular Formula Pt
Molecular Weight (g/mol) 195.08
MDL Number MFCD00011179
InChI Key BASFCYQUMIYNBI-UHFFFAOYSA-N
Synonym black, platin, sponge, platinum, metal, platinum, elemental, platine, platino, metal, platin german, iv ion
PubChem CID 23939
ChEBI CHEBI:33400
IUPAC Name platinum
SMILES [Pt]

Specifications

Quantity 5 g
UN Number UN1325
Merck Index 14,7529
Solubility Information Soluble in boiling aqua regia (reacts),and molten alkali cyanides (reacts). Insoluble in water,mineral acids,and organic acids.
Chemical Name or Material Platinum, 5% on carbon, Dry

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.