Learn More
Alfa Aesar™ Platinum, 5% on activated carbon paste, Type 18
Brand: Alfa Aesar™ H36968.14
Code : NEW
Additional Details : CAS Number : 7440-06-4 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
Platinum | |
7440-06-4 | |
5% on activated carbon paste, Type 18 | |
MFCD00011179 |
Specifications
Platinum | |
7440-06-4 | |
MFCD00011179 | |
Dark gray |
5% on activated carbon paste, Type 18 | |
Solid | |
Odorless | |
25g |