missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Platinum, 2% on 2.5mm alumina trilobes

Brand:  Thermo Scientific Alfa Aesar 045778.14

Platinum, 2% on 2.5mm alumina trilobes

Code : NEW

Additional Details : Weight : 0.02500kg

Product Code. 15482437

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.