Learn More
Platinum 10 wt% Rhodium wire, 0.25mm (0.01in) dia, ISA Type S Standard Grade Thermocouple, Thermo Scientific Chemicals
For ISA Type S Standard Grade Thermocouple
£582.00 - £2325.00
Specifications
Chemical Name or Material | Platinum 10 wt% Rhodium wire |
---|---|
Name Note | 0.25mm (0.01 in.) dia. |
Avg. Mol. Wt. or Mol. Wt. Range | Pt:Rh; 90:10 wt% |
MDL Number | MFCD00198186 |
Solubility Information | Insoluble in water. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316777
|
Thermo Scientific Alfa Aesar
010065.BU |
25 cm |
£582.00
25cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306777
|
Thermo Scientific Alfa Aesar
010065.BY |
100 cm |
£582.00
100cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15453495
|
Thermo Scientific Alfa Aesar
010065.CC |
500 cm |
£2325.00
500cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Platinum rhodium wire is used as a catalyst in preparation of nitric acid and in rayon spinnerets. It is used as a device for measuring high temperatures. It serves as a catalyst in the oxidation of the ammonia by atmospheric oxygen to produce nitric oxide, through Haber process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Platinum 10 wt% Rhodium wire | |
Pt:Rh; 90:10 wt% | |
Insoluble in water. | |
≈0.52g/50cm |
0.25mm (0.01 in.) dia. | |
MFCD00198186 | |
Odorless | |
Wire |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only