missing translation for 'onlineSavingsMsg'
Learn More

Platinum, 0.3% on 2.7-3.3mm (0.11-0.13in) alumina pellets, reduced, Thermo Scientific Chemicals

Product Code. 11349147
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349147 100 g 100g
11339147 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349147 Supplier Thermo Scientific Alfa Aesar Supplier No. 039826.22
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6-6-7440

Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Selective hydrogenation, gas purifications: e.g. Oxidation of carbon monoxide to carbon dioxide, hydrogen removal from oxygen or carbon dioxide
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6-6-7440
MDL Number MFCD00011179

Specifications

Quantity 100 g
Chemical Name or Material Platinum, 0.3% on 2.7 to 3.3mm (0.11 to 0.13 in.) alumina pellets, Reduced

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.