missing translation for 'onlineSavingsMsg'
Learn More

Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane Complex Solution, TRC

Product Code. 30413481
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30413481 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30413481

Brand: Toronto Research Chemicals TRCP578010250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Platinum(0)-1,3-divinyl-1,1,3,3-tetramethyldisiloxane Complex
CAS 68478-92-2
Molecular Formula C8H18OPtSi2
Quantity 250 mg
Synonym 1,1,3,3-Tetramethyl-1,3-divinyldisiloxane','Platinum Salt, Karstedt catalyst, Platinum','1,3-Diethenyl-1,1,3,3-tetramethyldisiloxane Complexes
InChI Formula InChI=1S/C8H18OSi2.Pt/c1-7-10(3,4)9-11(5,6)8-2;/h7-8H,1-2H2,3-6H3;
SMILES [Pt].C[Si](C)(O[Si](C)(C)C=C)C=C
IUPAC Name ethenyl-[ethenyl(dimethyl)silyl]oxy-dimethylsilane;platinum
Molecular Weight (g/mol) 381.48
Formula Weight 381.0544
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.