missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Platform trolley, steel, foldable, 300kg capacity
Description
Highly durable welded steel trolleys with a foldable handle. Non-slip PVC platform surface, ensures maximum grip of load when in transit. All round buffer strip protects walls and door frames when the truck is in use. Welded steel construction.
- Dimensions (W x L): 610mm x 920mm
- Handle height: 980mm
- Load capacity: 300kg
- Weight: 22.5kg
- Foot operated handle release bar
- Two fixed and two rear swivel non-marking tyres.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?