missing translation for 'onlineSavingsMsg'
Learn More

Key Industrial Equipment™ Platform trolley, steel, Ekwo, 150kg capacity

Brand:  Key Industrial Equipment™ A042426

Encompass_Preferred

Product Code. 12837438

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Dimensions (W x L): 480mm x 735mm
  • Fitted with two fixed and two rear swivel castors for easy manoeuvring.
  • Handle height: 820mm
  • Load capacity: 150kg
  • Weight: 8.2kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.