missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators

Accommodates tubes of different diameters. Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators keeps test tubes secure during rotation

Brand:  Grant Instruments™ PRS-10

Code : 04

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 11411078

  • £46.00 / Each


Estimated Shipment
14-02-2018
Add to basket

Description

Description

  • For use with PTR-35 and PTR-60 Series 360° vertical multi-function rotators
  • Hold as few as 8 test tubes or as many as 48.
  • Accommodates a variety of capacities
Specifications

Specifications

PTR-35
10 tubes up to 30mm, tube volumes up to 50mL
Videos