Learn More
Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators
Product Code. 11431028
Change view
Click to view available options
Holds:
10 Tubes up to 30 mm
14 tubes up to 30mm
22 Tubes up to 15 mm
26 Tubes up to 15 mm, 1.5 -15 mL
48 Tubes up to 15 mm
5 x 30 mm and 12 x 15 mm tubes
8 x 30 mm, 22 x 15 mm tubes
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
11431028 22 Tubes up to 15 mm Each
11411078 10 Tubes up to 30 mm Each
11421028 14 tubes up to 30mm Each
11491068 26 Tubes up to 15 mm, 1.5 -15 mL Each
11411028 48 Tubes up to 15 mm Each
10860891 5 x 30 mm and 12 x 15 mm tubes Each
12915511 8 x 30 mm, 22 x 15 mm tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11431028 Supplier Grant Instruments™ Supplier No. PRS22
Only null left
Add to basket
Add to basket
Accommodates tubes of different diameters. Grant Instruments™ Platform for PTR 35 and 60 Series Compact Variable Speed / Angle Vertical Rotators keeps test tubes secure during rotation
  • For use with PTR-35 and PTR-60 Series 360° vertical multi-function rotators
  • Hold as few as 8 test tubes or as many as 48.
  • Accommodates a variety of capacities
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) PTR-30
Holds 22 Tubes up to 15 mm