Learn More
Thermo Scientific™ Accessories for Digital Microplate Shakers
Product Code. 16504802 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
96-well microplate clamps
Platform Large
Platform Large Assemble
Dimensions (L x W x H):
54 x 19 x 19 mm
292 x 275 x 14 mm
292 x 275 x 25 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Dimensions (L x W x H) unitSize
16504802 96-well microplate clamps 54 x 19 x 19 mm Each
16594792 Platform Large 292 x 275 x 14 mm Each
16514802 Platform Large Assemble 292 x 275 x 25 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16504802 Supplier Thermo Scientific™ Supplier No. 88882115
Only null left
Add to basket
Add to basket
Includes
Pack of 7 Clamps
Get assorted accessories for Digital Microplate Shakers
Assorted accessories for Digital Microplate Shakers
Service Plan
Standard Thermo warranty guideline, provide repair service and free spare part within warranty
Warranty
2 years
TRUSTED_SUSTAINABILITY
Specifications
Product Type 96-well Microplate Clamps
Material Stainless Steel
Dimensions (L x W x H) 54 x 19 x 19 mm
Description 96-well microplate clamps
For Use With (Equipment) Rocker, Rotator, mixer
Includes 7 Clamps