Learn More
Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces
Help reduce variability in molecular orientation with these plates and modules with affinity binding surfaces.
Brand:  Thermo Scientific™ 95029263
Code : 04
Additional Details : Weight : 0.34700kg
 View more versions of this product
Product Code. 10723686
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Help reduce variability in molecular orientation with Thermo Scientific™ Plates and Modules with Affinity Binding Surfaces.
Affinity Capture Surfaces (Passive Streptavidin and Immobilizer Streptavidin, Nickel Chelate and Glutathione, BioBind)
  • Highly specific binding
  • Reduced variability in molecular orientation
  • Immobilizer surfaces improve signal to noise ratios
  • Streptavidin biotin interaction can be exploited to immobilize a wide range of biomolecules
Specifications
Specifications
Colorimetric
Affinity
Microplate
Streptavidin
Flat Bottom (F-well)
5
Biotinylated
5 Cases
Clear
200 μL
400 μL
5
Unspecified